Få mere at vide
(3Beta,5Alpha,6Alpha,20R)-Pregnane-3,6,20-triol, TRC
Artikelnummer. 30409038
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409038 10 mg
30409405 2.5 mg
30416676 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30409038 Leverandør Toronto Research Chemicals Leverandørnr. TRCP03099010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70286-67-8
Molekylær formel C21H36O3
Molekylvægt (g/mol) 336.51
Synonym 5a-Pregane-3b,6a,20b-triol
IUPAC navn (3S,5S,6S,8S,9S,10R,13S,14S,17S)-17-[(1R)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,6-diol
SMIL C[C@@H](O)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale (3beta,5alpha,6alpha,20R)-Pregnane-3,6,20-triol
Mængde 10 mg
InChI formel InChI=1S/C21H36O3/c1-12(22)15-4-5-16-14-11-19(24)18-10-13(23)6-8-21(18,3)17(14)7-9-20(15,16)2/h12-19,22-24H,4-11H2,1-3H3/t12-,13+,14+,15-,16+,17+,18-,19+,20-,21-/m1/s1
Formel vægt 336.2664