Få mere at vide
3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one, Mikromol™
Artikelnummer. 30050045
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050045 100 mg
30050345 25 mg
30462384 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050045 Leverandør Mikromol Leverandørnr. MM1203.12
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one
CAS 82543-15-5
Analyt- eller komponentnavne 3beta,5-Dihydroxy-15beta,16beta-methylene-5beta-androst-6-en-17-one
Forsendelsestilstand Room Temperature
Molekylær formel C20 H28 O3
Certificeringer/compliance ISO 17025
Synonym 3'H-Cycloprop[15,16]androsta-6,15-dien-17-one, 15,16-dihydro-3,5-dihydroxy-, (3β,5β,15α,16α)-, 3β,5-Dihydroxy-15β,16β-methylene-5β-androst-6-en-17-one
InChI formel InChI=1S/C20H28O3/c1-18-6-5-15-12(16(18)13-9-14(13)17(18)22)4-8-20(23)10-11(21)3-7-19(15,20)2/h4,8,11-16,21,23H,3,5-7,9-10H2,1-2H3/t11-,12+,13+,14-,15-,16+,18-,19+,20-/m0/s1
SMIL C[C@]12CC[C@H]3[C@@H](C=C[C@]4(O)C[C@@H](O)CC[C@]34C)[C@@H]1[C@@H]5C[C@@H]5C2=O
Molekylvægt (g/mol) 316.43
Mængde 100 mg
Formel vægt 316.2038 g/mol