Få mere at vide
(3Beta)-3-(3-Azidopropoxy)solanid-5-ene, TRC
Artikelnummer. 30425540
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30425540 5 mg -
30424615 2.5 mg -
30417076 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30425540 Leverandør Toronto Research Chemicals Leverandørnr. TRCS0645505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1644551-84-7
Molekylær formel C30H48N4O
Molekylvægt (g/mol) 480.73
Synonym (4S,6aR,6bS,8aS,8bR,9S,9aR,12S,14aS,15aS,15bS)-4-(3-Azidopropoxy)-6a,8a,9,12-tetramethyl-3,4,5,6,6a,6b,7,8,8a,8b,9,9a,10,11,12,13,14a,15,15a,15b-icosahydro-1H-naphtho[2',1':4,5]indeno[1,2-b]indolizine
SMIL C[C@H]1CC[C@@H]2[C@@H](C)[C@H]3[C@H](C[C@H]4[C@@H]5CC=C6C[C@H](CC[C@]6(C)[C@H]5CC[C@]34C)OCCCN=[N+]=[N-])N2C1
Tekniske data
Kemisk navn eller materiale (3beta)-3-(3-Azidopropoxy)solanid-5-ene
Mængde 5 mg
InChI formel InChI=1S/C30H48N4O/c1-19-6-9-26-20(2)28-27(34(26)18-19)17-25-23-8-7-21-16-22(35-15-5-14-32-33-31)10-12-29(21,3)24(23)11-13-30(25,28)4/h7,19-20,22-28H,5-6,8-18H2,1-4H3/t19-,20+,22-,23+,24-,25-,26+,27-,28-,29-,30-/m0/s1
Formel vægt 480.3828