missing translation for 'onlineSavingsMsg'
Få mere at vide

(3Beta,20R)-Pregn-5-ene-3,17,20-triol, TRC

Artikelnummer. 30420173
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420173 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30420173 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7123205MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (3beta,20R)-Pregn-5-ene-3,17,20-triol
CAS 2204-13-9
Molekylær formel C21H34O3
Mængde 5 mg
Synonym Pregn-5-ene-3beta,17,20beta-triol, 3beta,17alpha,20beta-Trihydroxypregn-5-ene
InChI formel InChI=1S/C21H34O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,13,15-18,22-24H,5-12H2,1-3H3/t13-,15+,16-,17+,18+,19+,20+,21+/m1/s1
SMIL C[C@@H](O)[C@@]1(O)CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (3S,8R,9S,10R,13S,14S,17R)-17-[(1R)-1-hydroxyethyl]-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 334.49
Formel vægt 334.2508
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.