Få mere at vide
(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one (Mutilin), Mikromol™
Artikelnummer. 30051030
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051030 25 mg
17914006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051030 Leverandør Mikromol Leverandørnr. MM0394.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one (Mutilin)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one (Mutilin)
CAS 6040-37-5
Analyt- eller komponentnavne (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one (Mutilin)
Forsendelsestilstand Room Temperature
Molekylær formel C20 H32 O3
Certificeringer/compliance ISO 17025
Synonym Tiamulin Hydrogen Fumarate Imp. A (EP), Tiamulin Imp. A (EP), (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one, Mutilin, Tiamulin Hydrogen Fumarate Impurity A, Tiamulin Impurity A
InChI formel InChI=1S/C20H32O3/c1-6-18(4)11-15(22)19(5)12(2)7-9-20(13(3)17(18)23)10-8-14(21)16(19)20/h6,12-13,15-17,22-23H,1,7-11H2,2-5H3/t12-,13+,15-,16+,17+,18-,19+,20+/m1/s1
SMIL C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@H](O)C[C@@](C)(C=C)[C@@H](O)[C@@H]3C
IUPAC navn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5,8-dihydroxy-4,6,9,10-tetramethyloctahydro-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one
Molekylvægt (g/mol) 320.47
Mængde 25 mg
Formel vægt 320.2351 g/mol