Få mere at vide
(3aR,4R,5R,6aS)-4-[(E,3S)-3-Hydroxy-5-phenylpent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol, Mikromol™
Artikelnummer. 30052631
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052631 100 mg
30049385 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052631 Leverandør Mikromol Leverandørnr. MM3316.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3aR,4R,5R,6aS)-4-[(E,3S)-3-Hydroxy-5-phenylpent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3aR,4R,5R,6aS)-4-[(E,3S)-3-Hydroxy-5-phenylpent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol
CAS 856240-62-5
Analyt- eller komponentnavne (3aR,4R,5R,6aS)-4-[(E,3S)-3-Hydroxy-5-phenylpent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol
Forsendelsestilstand Room Temperature
Molekylær formel C18 H24 O4
Certificeringer/compliance ISO 17025
Synonym 2H-Cyclopenta[b]furan-2,5-diol, hexahydro-4-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]-, (3aR,4R,5R,6aS)-, 2H-Cyclopenta[b]furan-2,5-diol, hexahydro-4-[(1E,3S)-3-hydroxy-5-phenyl-1-pentenyl]-, (3aR,4R,5R,6aS)-, (3aR,4R,5R,6aS)-4-[(E,3S)-3-Hydroxy-5-phenyl-pent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol
InChI formel InChI=1S/C18H24O4/c19-13(7-6-12-4-2-1-3-5-12)8-9-14-15-10-18(21)22-17(15)11-16(14)20/h1-5,8-9,13-21H,6-7,10-11H2/b9-8+/t13-,14+,15+,16+,17-,18?/m0/s1
SMIL O[C@@H](CCc1ccccc1)\C=C\[C@H]2[C@H](O)C[C@@H]3OC(O)C[C@H]23
IUPAC navn (3aR,4R,5R,6aS)-4-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]furan-2,5-diol
Molekylvægt (g/mol) 304.38
Mængde 100 mg
Formel vægt 304.1675 g/mol