Få mere at vide
3Alpha,7Alpha-Dihydroxycoprostanic Acid-d3, TRC
Artikelnummer. 30369930
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30369930 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369930 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4525170.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3alpha,7alpha-Dihydroxycoprostanic Acid-d3
CAS 338976-76-4
Molekylær formel C27H43D3O4
Mængde 0.5 mg
Synonym (3α,5β,7α)-3,7-Dihydroxycholestan-26-oic Acid-d3, 3α,7α-Dihydroxy-5β-cholestan-26-oic Acid-d3, 3α,7α-Dihydroxy-5β-cholestanoic Acid-d3, 3α,7α-Hydroxy-5β-cholestan-26-oic Acid-d3, (3α,5β,7α)-3,7-Dihydroxy-cholestan-26-oic-27,27,27-d3 Acid
InChI formel InChI=1S/C27H46O4/c1-16(6-5-7-17(2)25(30)31)20-8-9-21-24-22(11-13-27(20,21)4)26(3)12-10-19(28)14-18(26)15-23(24)29/h16-24,28-29H,5-15H2,1-4H3,(H,30,31)/t16-,17?,18+,19-,20-,21+,22+,23-,24+,26+,27-/m1/s1/i2D3
SMIL O[C@@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@]3(CC[C@@]4([C@H](C)CCCC(C([2H])([2H])[2H])C(O)=O)[H])[H])([H])C1
Molekylvægt (g/mol) 437.67