Få mere at vide
3Alpha,7Alpha,12Alpha-Trihydroxycoprostanic Acid-d3, TRC
Artikelnummer. 30450223
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30450223 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450223 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79515710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3alpha,7alpha,12alpha-Trihydroxycoprostanic Acid-d3
CAS 338976-79-7
Molekylær formel C27H43D3O5
Mængde 10 mg
Synonym (3α,5β,7α,12α)-3,7,12-Trihydroxy-cholestan-26-oic-27,27,27-d3 Acid, (3α,5β,7α,12α)-3,7,12-Trihydroxycholestan-26-oic Acid-d3, 3α,7α,12α-Trihydroxy-5β-cholestan-26-oic Acid-d3, 3α,7α,12α-Trihydroxycoprostanic Acid-d3, 5β-Cholestane-3α,7α,12α-triol-26-oic Acid-d3, Coprocholic Acid-d3, Trihydroxycoprostanic Acid-d3
InChI formel InChI=1S/C27H46O5/c1-15(6-5-7-16(2)25(31)32)19-8-9-20-24-21(14-23(30)27(19,20)4)26(3)11-10-18(28)12-17(26)13-22(24)29/h15-24,28-30H,5-14H2,1-4H3,(H,31,32)/t15-,16?,17+,18-,19-,20+,21+,22-,23+,24+,26+,27-/m1/s1/i2D3
SMIL O[C@@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@]3(CC[C@@]4([C@H](C)CCCC(C([2H])([2H])[2H])C(O)=O)[H])[H])([H])C1
Molekylvægt (g/mol) 453.67
Formel vægt 453.3534