Få mere at vide
(3Alpha,5Beta,7Alpha,12Alpha)-3,7,12-Trihydroxycholan-2,2,3,4,4-d5-24-oic Acid Methyl Ester, TRC
Artikelnummer. 30476581
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30476581 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476581 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2944772.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (3Alpha,5Beta,7Alpha,12Alpha)-3,7,12-Trihydroxycholan-2,2,3,4,4-d5-24-oic Acid Methyl Ester
CAS 52886-39-2
Molekylær formel C25H37D5O5
Mængde 2.5 mg
Synonym Methyl Cholate-d5, Cholic-d5 Acid Methyl Ester, Methyl 3α,7α,12α-Trihydroxy-5β-cholan-24-ate-d5, Methyl 3α,7α,12α-Trihydroxy-5β-cholan-24-oate-d5, Methyl 3α,7α,12α-Trihydroxy-5β-cholanoate-d5
InChI formel InChI=1S/C25H42O5/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-21,23,26-28H,5-13H2,1-4H3/t14-,15+,16-,17-,18+,19+,20-,21+,23+,24+,25-/m1/s1/i9D2,11D2,16D
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3C[C@H](O)[C@]4(C)[C@H](CC[C@H]4[C@@H]3[C@H](O)C[C@@H]2C([2H])([2H])[C@]1([2H])O)[C@H](C)CCC(=O)OC
Molekylvægt (g/mol) 427.63