Få mere at vide
(3alpha,5alpha)-3,17-Dihydroxypregnan-20-one, TRC
Artikelnummer. 30012381
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012381 100 mg
30012577 50 mg
30010564 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012381 Leverandør Toronto Research Chemicals Leverandørnr. TRCD863770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6890-65-9
Molekylær formel C21 H34 O3
Molekylvægt (g/mol) 334.493
Synonym 1-((3R,3aS,6aS,6bS,9R,10aS,12aS,13bS)-3,9-dihydroxy-3a,6b-dimethylicosahydro-1H-cyclopenta[k]tetraphen-3-yl)ethanone;
IUPAC navn 1-[(3R,5S,8R,9S,10S,13S,14S,17R)-3,17-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
SMIL CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale (3alpha,5alpha)-3,17-Dihydroxypregnan-20-one
Mængde 100 mg
InChI formel InChI=1S/C21H34O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h14-18,23-24H,4-12H2,1-3H3/t14-,15+,16+,17-,18-,19-,20-,21-/m0/s1
Formel vægt 334.251