Få mere at vide
3a,6,7,9a-Tetrahydro-1,3,2-dioxathiolo[4,5-c]azocine-5(4H)-carboxylic Acid Phenylmethyl Ester 2-Oxide, TRC
Artikelnummer. 30411422
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411422 5 mg
30425177 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30411422 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2935655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3a,6,7,9a-Tetrahydro-1,3,2-dioxathiolo[4,5-c]azocine-5(4H)-carboxylic Acid Phenylmethyl Ester 2-Oxide
CAS 1315552-02-3
Molekylær formel C15H17NO5S
Mængde 5 mg
InChI formel InChI=1S/C15H17NO5S/c17-15(19-11-12-6-2-1-3-7-12)16-9-5-4-8-13-14(10-16)21-22(18)20-13/h1-4,6-8,13-14H,5,9-11H2/b8-4-/t13-,14+,22?/m0/s1
SMIL [H][C@@]12[C@@](OS(O2)=O)([H])CN(C(OCC3=CC=CC=C3)=O)CC/C=C\1
IUPAC navn benzyl (3aR,9aS,Z)-3a,6,7,9a-tetrahydro-[1,3,2]dioxathiolo[4,5-c]azocine-5(4H)-carboxylate 2-oxide
Molekylvægt (g/mol) 323.36
Formel vægt 323.08