Få mere at vide
3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-(methyl)-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol-d3, TRC
Artikelnummer. 30419807
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30419807 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419807 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2936272.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-(methyl)-(3aR,5R,6S,7R,7aR)-5H-Pyrano[3,2-d]thiazole-6,7-diol-d3
CAS 944070-25-1
Molekylær formel C8 D3 H10 N O4 S
Mængde 2.5 mg
InChI formel InChI=1S/C8H13NO4S/c1-3-9-5-7(12)6(11)4(2-10)13-8(5)14-3/h4-8,10-12H,2H2,1H3/t4-,5-,6-,7?,8-/m1/s1/i1D3
SMIL [2H]C([2H])([2H])C1=N[C@H]2[C@H](O[C@H](CO)[C@@H](O)C2O)S1
IUPAC navn (3aR,5R,6S,7aR)-5-(hydroxymethyl)-2-(trideuteriomethyl)-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
Molekylvægt (g/mol) 222.277
Formel vægt 222.075