Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Brand: RETSCH 02.407.0033
2451.00 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.15000kg
Specifications
1Each | |
Ring Sieves | |
SR200/SR300 Rotor Beater Mills |
1.5mm | |
Stainless Steel |