Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Brand:  RETSCH 02.407.0032
13713.25 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 10354241
  • 14435.00 DKK / Each
Estimated Shipment: 24-07-2024
to see stock.
Specifications
1Each
Ring Sieves
SR200/SR300 Rotor Beater Mills
1mm