Learn More
Sampling Systems™ 316 Stainless Steel Storage Containers without Toggle Clamps
Multi-purpose containers with light duty construction for a wide range of storage uses in the laboratory.
Brand: Sampling Systems™ A466-100
Additional Details : Weight : 30.92600kg
Description
- Crevice-free interior
- Two sturdy handles
- Close fitting recessed lid with handle
- Flat bottom with chime ring to give added protection
- Toggle clamps and seals are optional extras
Specifications
Storage container | |
Recessed Lid | |
Stainless Steel | |
445 mm |
Yes | |
100 L | |
675 mm |