Få mere at vide
Lactase Antibody (CL14164) [DyLight 350], Novus Biologicals Biologicals™
Shop alle Bio Techne produkterBeskrivelse
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antigen | Lactase |
| Applikationer | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL14164 |
| Konjugeret | DyLight 350 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 |
| Værtsarter | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of P09848, with the exact immunogen sequence remaining proprietary. |
| Oprensningsmetode | Protein A purified |
| Vis mere |