Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve with Square Holes
Brand: RETSCH 60.138.014000
1957.00 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.50000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel |
For separation, fractioning, particle size determination | |
305 mm | |
14mm | |
ISO 3310/2 | |
40mm |