Få mere at vide
CBFA2T3 Antibody [Allophycocyanin], Novus Biologicals Biologicals™
Artikelnummer. 30499071 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Artikelnummer. Mængde
30499071 0.1 mL
1 options
Artikelnummer. 30499071 Leverandør Novus Biologicals Leverandørnr. NBP335460APC
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
CBFA2T3 Polyclonal antibody specifically detects CBFA2T3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen CBFA2T3
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret APC
Formulering PBS
Gene Alias core-binding factor, runt domain, alpha subunit 2; translocated to, 3, hMTG16, MTG16ETO2, MTG8-related protein 2, MTGR2MTG8-related gene 2, Myeloid translocation gene on chromosome 16 protein, protein CBFA2T3, Zinc finger MYND domain-containing protein 4, ZMYND4myeloid translocation gene 8 and 16b
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 593-653 of human CBFA2T3 (NP_005178.4).,, Sequence:, DRDPLHPEHLSKRPCTLNPAQRYSPSNGPPQPTPPPHYRLEDIAMAHHFRDAYRHPDPRELRERHRPLVVPGSRQEEVIDHKLTER
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 863
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG