missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
TCAP Polyclonal antibody specifically detects TCAP in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
Specifications
| Antigen | TCAP |
| Applikationer | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Alexa Fluor 700 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | 19 kDa sarcomeric protein, CMD1NTitin cap protein, LGMD2G, limb girdle muscular dystrophy 2G (autosomal recessive), T-cap, TELE, telethonin, titin-cap (telethonin) |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human TCAP (NP_003664.1).,, Sequence:, MATSELSCEVSEENCERREAFWAEWKDLTLSTRPEEGCSLHEEDTQRHETYHQQGQCQVLVQRSPWLMMRMGILGRGLQEYQLPYQRVLPLPIFTPAKMGATKEEREDTPIQLQELLALETALGGQCVDRQEVAEITKQLPPVVPVSKPGALRRSLSRSMSQEAQRG |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?