missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
SMARCD2 Polyclonal antibody specifically detects SMARCD2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antigen | SMARCD2 |
| Applikationer | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 650 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | 60 kDa BRG-1/Brm-associated factor subunit B, BAF60B, BRG1-associated factor 60B, chromatin remodeling complex BAF60B subunit, CRACD2, mammalian chromatin remodeling complex BRG1-associated factor 60B, PRO2451, Rsc6p, SWI/SNF complex 60 kDa subunit B, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2, Swp73-like protein |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 454-531 of human SMARCD2 (NP_001091896.1).,, Sequence:, RDFMLSFSTDPQDFIQEWLRSQRRDLKIITDVIGNPEEERRAAFYHQPWAQEAVGRHIFAKVQQRRQELEQVLGIRLT |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?