Få mere at vide
DLC1 Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™
Artikelnummer. 30498967 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30498967 0.1 mL 0.1ml
1 options
Artikelnummer. 30498967 Leverandør Novus Biologicals Leverandørnr. NBP335894AF700
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
DLC1 Polyclonal antibody specifically detects DLC1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen DLC1
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret Alexa Fluor 700
Formulering 50mM Sodium Borate
Gene Alias ARHGAP7 STARD12 deleted in liver cancer 1 variant 2, deleted in liver cancer 1, Deleted in liver cancer 1 protein, DLC-1, FLJ21120, HP, HP protein, KIAA1723, p122-RhoGAP, rho GTPase-activating protein 7, Rho-GTPase-activating protein 7, Rho-type GTPase-activating protein 7, StARD12, StAR-related lipid transfer (START) domain containing 12, StAR-related lipid transfer protein 12, START domain-containing protein 12
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 204-463 of human DLC1 (NP_079043.3).,, Sequence:, DAPKVNAVDTLNVKDIAPEKQLLNSAVIAQQRRKPDPPKDENERSTCNVVQNEFLDTPCTNRGLPLLKTDFGSCLLQPPSCPNGMSAENGLEKSGFSQHQNKSPPKVKAEDGMQCLQLKETLATQEPTDNQVRLRKRKEIREDRDRARLDSMVLLIMKLDQLDQDIENALSTSSSPSGTPTNLRRHVPDLESGSESGADTISVNQTRVNLSSDTESTDLPSSTPVANSGTKPKTTAIQGISEKEKAGKLTFWFCFLANLF
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 10395
Målarter Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG