missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Description
WDR61 Polyclonal antibody specifically detects WDR61 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | WDR61 |
| Applikationer | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 755 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | Meiotic recombination REC14 protein homolog, REC14, recombination protein REC14, SKI8, WD repeat domain 61, WD repeat-containing protein 61 |
| Værtsarter | Rabbit |
| Immunogen | WDR61 antibody was raised against a 14 amino acid peptide near the amino terminus of human WDR61. The immunogen is located within the first 50 amino acids of WDR61. |
| Oprensningsmetode | Peptide affinity purified |
| Mængde | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?