missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
SMNDC1 Polyclonal antibody specifically detects SMNDC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Tekniske data
Tekniske data
| Antigen | SMNDC1 |
| Applikationer | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 350 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30 |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-238 of human SMNDC1 (NP_005862.1).,, Sequence:, MSEDLAKQLASYKAQLQQVEAALSGNGENEDLLKLKKDLQEVIELTKDLLSTQPSETLASSDSFASTQPTHSWKVGDKCMAVWSEDGQCYEAEIEEIDEENGTAAITFAGYGNAEVTPLLNLKPVEEGRKAKEDSGNKPMSKKEMIAQQREYKKKKALKKAQRIKELEQEREDQKVKWQQFNNRAYSKNKKGQVKRSIFASPESVTGKVGVGTCGIADKPMTQYQDTSKYNVRHLMPQ |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?