missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
RAPGEF3 Polyclonal antibody specifically detects RAPGEF3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Tekniske data
Tekniske data
| Antigen | RAPGEF3 |
| Applikationer | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | Alexa Fluor 700 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | 9330170P05Rik, bcm910, cAMP-GEFI, cAMP-regulated guanine nucleotide exchange factor I, CGEF1, EPAC 1, EPAC1, EPACRAP guanine-nucleotide-exchange factor (GEF) 3, Exchange factor directly activated by cAMP 1, Exchange protein directly activated by cAMP 1, HSU79275, MGC21410, Rap guanine nucleotide exchange factor (GEF) 3, rap guanine nucleotide exchange factor 3, Rap1 guanine-nucleotide-exchange factor directly activated by cAMP |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAPGEF3 (NP_001092002.1).,, Sequence:, MVLRRMHRPRSCSYQLLLEHQRPSCIQGLRWTPLTNSEESLDFSESLEQASTERVLRAGRQLHRHLLATCPNLIRDRKYHLRLYRQCCSGRELVDGILALGLGVHSRSQVVGICQVLLDEGALCHVKHDWAFQDRDAQFYRFPGPEPEPVGTHEMEEELAEAVALLSQRGPDALLTVALRKPPGQRTDEELDLIFEELLH |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?