Få mere at vide
Proteasome 20S alpha 6 Antibody [PerCP], Novus Biologicals Biologicals™
Artikelnummer. 30498906 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30498906 0.1 mL 0.1ml
1 options
Artikelnummer. 30498906 Leverandør Novus Biologicals Leverandørnr. NBP335356PCP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
Proteasome 20S alpha 6 Polyclonal antibody specifically detects Proteasome 20S alpha 6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen Proteasome 20S alpha 6
Applikationer ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Klassifikation Polyclonal
Konjugeret PerCP
Formulering PBS
Gene Alias EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-246 of human Proteasome 20S alpha 6 (NP_002782.1).,, Sequence:, MSRGSSAGFDRHITIFSPEGRLYQVEYAFKAINQGGLTSVAVRGKDCAVIVTQKKVPDKLLDSSTVTHLFKITENIGCVMTGMTADSRSQVQRARYEAANWKYKYGYEIPVDMLCKRIADISQVYTQNAEMRPLGCCMILIGIDEEQGPQVYKCDPAGYYCGFKATAAGVKQTESTSFLEKKVKKKFDWTFEQTVETAITCLSTVLSIDFKPSEIEVGVVTVENPKFRILTEAEIDAHLVALAERD
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5687
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG