Beskrivelse
EPLIN Polyclonal antibody specifically detects EPLIN in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Tekniske data
Tekniske data
| Antigen | EPLIN |
| Applikationer | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 405 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3 |
| Værtsarter | Rabbit |
| Immunogen | EPLIN antibody was raised against an 18 amino acid peptide near the amino terminus of human EPLIN. The immunogen is located within amino acids 230 - 280 of EPLIN. |
| Oprensningsmetode | Peptide affinity purified |
| Mængde | 0.1 mL |
| Vis mere |