Få mere at vide
NRAMP2/SLC11A2/DMT1 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™
Shop alle Bio Techne produkterBeskrivelse
NRAMP2/SLC11A2/DMT1 Polyclonal antibody specifically detects NRAMP2/SLC11A2/DMT1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antigen | NRAMP2/SLC11A2/DMT1 |
| Applikationer | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugeret | Alexa Fluor 405 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | DCT1NRAMP 2, Divalent cation transporter 1, Divalent metal transporter 1, DMT-1, DMT1FLJ37416, member 2, NRAMP2natural resistance-associated macrophage protein 2, solute carrier family 11 (proton-coupled divalent metal ion transporters) |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-63 of human NRAMP2/SLC11A2/DMT1 (NP_001167597.1).,, Sequence:, MVLGPEQKMSDDSVSGDHGESASLGNINPAYSNPSLSQSPGDSEEYFATYFNEKISIPEEEYS |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |