missing translation for 'onlineSavingsMsg'
Få mere at vide

DHX38 Antibody [DyLight 550], Novus Biologicals Biologicals™

Artikelnummer. 30498825 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30498825 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30498825 Leverandør Novus Biologicals Leverandørnr. NBP338087R

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

DHX38 Polyclonal antibody specifically detects DHX38 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX38
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret DyLight 550
Formulering 50mM Sodium Borate
Gene Alias ATP-dependent RNA helicase DHX38, DDX38pre-mRNA-splicing factor ATP-dependent RNA helicase PRP16, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 38, DEAH (Asp-Glu-Ala-His) box polypeptide 38, DEAH box protein 38, EC 3.6.1, EC 3.6.4.13, KIAA0224pre-mRNA splicing factor ATP-dependent RNA helicase PRP16, PRP16 homolog of S.cerevisiae, PRP16hPrp16, PRPF16
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1133-1227 of human DHX38 (NP_054722.2).,, Sequence:, VTAVDGEWLAELGPMFYSVKQAGKSRQENRRRAKEEASAMEEEMALAEEQLRARRQEQEKRSPLGSVRSTKIYTPGRKEQGEPMTPRRTPARFGL
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 9785
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.