missing translation for 'onlineSavingsMsg'
Få mere at vide

SUG1 Antibody [DyLight 550], Novus Biologicals Biologicals™

Artikelnummer. 30498705 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30498705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30498705 Leverandør Novus Biologicals Leverandørnr. NBP335357R

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

SUG1 Polyclonal antibody specifically detects SUG1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SUG1
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret DyLight 550
Formulering 50mM Sodium Borate
Gene Alias MSUG1 protein, p45, p45/SUG26S protease regulatory subunit 8, proteasome (prosome, macropain) 26S subunit, ATPase, 5, proteasome 26S ATPase subunit 5, Proteasome 26S subunit ATPase 5, Proteasome subunit p45, S8, SUG-1, SUG126S proteasome AAA-ATPase subunit RPT6, Tat-binding protein homolog 10, TBP10, thyroid receptor interactor 1, TRIP1Thyroid hormone receptor-interacting protein 1
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 127-406 of human SUG1 (NP_002796.4).,, Sequence:, LPNKVDPLVSLMMVEKVPDSTYEMIGGLDKQIKEIKEVIELPVKHPELFEALGIAQPKGVLLYGPPGTGKTLLARAVAHHTDCTFIRVSGSELVQKFIGEGARMVRELFVMAREHAPSIIFMDEIDSIGSSRLEGGSGGDSEVQRTMLELLNQLDGFEATKNIKVIMATNRIDILDSALLRPGRIDRKIEFPPPNEEARLDILKIHSRKMNLTRGINLRKIAELMPGASGAEVKGVCTEAGMYALRERRVHVTQEDFEMAVAKVMQKDSEKNMSIKKLWK
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5705
Målarter Human, Mouse
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.