missing translation for 'onlineSavingsMsg'
Få mere at vide

CRMP1 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Artikelnummer. 30498638 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30498638 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30498638 Leverandør Novus Biologicals Leverandørnr. NBP338009AF647

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

CRMP1 Polyclonal antibody specifically detects CRMP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CRMP1
Applikationer ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Klassifikation Polyclonal
Konjugeret Alexa Fluor 647
Formulering 50mM Sodium Borate
Gene Alias collapsin response mediator protein 1DRP1, CRMP-1, dihydropyrimidinase-like 1, dihydropyrimidinase-related protein 1, DPYSL1DRP-1dihydropyrimidinase related protein-1, ULIP3, ULIP-3, Unc-33-like phosphoprotein 3
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 460-540 of human CRMP1 (NP_001304.1).,, Sequence:, NVNKGMGRFIPRKAFPEHLYQRVKIRNKVFGLQGVSRGMYDGPVYEVPATPKYATPAPSAKSSPSKHQPPPIRNLHQSNFS
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Neuroscience
Primær eller sekundær Primary
Gen-id (Entrez) 1400
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.