missing translation for 'onlineSavingsMsg'
Få mere at vide

Cytochrome P450 2D6 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™

Artikelnummer. 30498256 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30498256 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30498256 Leverandør Novus Biologicals Leverandørnr. NBP335296AF405

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Cytochrome P450 2D6 Polyclonal antibody specifically detects Cytochrome P450 2D6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Cytochrome P450 2D6
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret Alexa Fluor 405
Formulering 50mM Sodium Borate
Gene Alias CPD6P450DB1, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, cytochrome P450 2D6, cytochrome P450, family 2, subfamily D, polypeptide 6, cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2, cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2, cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), Cytochrome P450-DB1, Debrisoquine 4-hydroxylase, EC 1.14.14.1, flavoprotein-linked monooxygenase, -metabolizing)-like 1, MGC120389, MGC120390, microsomal monooxygenase, P450C2D, P450-DB1, polypeptide 6, polypeptide 7 pseudogene 2, polypeptide 8 pseudogene 2, xenobiotic monooxygenase
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human Cytochrome P450 2D6 (NP_000097.3).,, Sequence:, VDLMHRRQRWAARYPPGPLPLPGLGNLLHVDFQNTPYCFDQLRRRFGDVFSLQLAWTPVVVLNGLAAVREALVTHGEDTADRPPVPITQILGFGPRSQGVFLARYGPAWREQRRFSVSTLRNLGLGKKSLEQWVTEEAACLCAAFANHSGRPFRPNGLLDKAVSNVIASLTCGRRFEYDDPRFLRLLDLAQEGLKEESGFLREVLNAVPVL
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primær eller sekundær Primary
Gen-id (Entrez) 1565
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.