Få mere at vide
kinectin Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Artikelnummer. 30497782 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30497782 0.1 mL 0.1ml
1 options
Artikelnummer. 30497782 Leverandør Novus Biologicals Leverandørnr. NBP336718MFV610
Rabbit Polyclonal Antibody
kinectin Polyclonal antibody specifically detects kinectin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen kinectin
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret mFluor Violet 610 SE
Formulering 50mM Sodium Borate
Gene Alias kinectin, kinectin 1 (kinesin receptor), MGC133337
Værtsarter Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human kinectin (NP_001072989.1).,, Sequence:, MEFYESAYFIVLIPSIVITVIFLFFWLFMKETLYDEVLAKQKREQKLIPTKTDKKKAEKKKNKKKEIQNGNLHESDSESVPRDFKLSDALAVEDDQVAPV
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 3895
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG