Learn More
Nickel Electro™ Stainless Steel Laboratory Funnel
Product Code. 12973621 Shop All Nickel-Electro Products
Change view
Click to view available options
Diameter (Metric)Top:
103 mm
165 mm
210 mm
Length (Metric) Stem:
55 mm
75 mm
90 mm
Height (Metric):
115 mm
158 mm
205 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Length (Metric) Stem Height (Metric) Unit Size
12973621 165 mm 75 mm 158 mm Each
12963621 103 mm 55 mm 115 mm Each
12983621 210 mm 90 mm 205 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12973621 Supplier Nickel Electro™ Supplier No. FUNSS150H
  • ONLINE EXCLUSIVE PRICE
    209.00 DKK
    182.29 DKK / Pakke med 1
    Spare 26.71 DKK
    12% Slukket
Forventet forsendelse: 02-02-2027
Use with or without filter membranes for gravity filtration as well as simple funnel procedures. Nickel Electro™ Stainless Steel Laboratory Funnel is vented to allow complete, uninhibited flow of liquids or solids.
  • Durable stainless steel funnel, break-resistant, stain proof and easy to clean.
  • May be used with or without filter membranes for gravity filtration as well as simple funnel procedures.
  • Flute is vented to allow complete, uninhibited flow.
  • Can be used with liquids or solids.
  • The handle can also be used as a convenient hanging point when not in use.
  • Smooth finish on internal surfaces making funnel easy to clean or sterilise.
Specifications
Diameter (Metric)Top 165 mm
Material Stainless Steel
Height (Metric) 158 mm
Includes Handle
Length (Metric) Stem 75 mm