Learn More
Gilson™ 3-Way Valve Kit
Product Code. 16686135 Shop All Gilson Products
Change view
Click to view available options
Description:
FC203
FC204
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
16686135 FC203 Each
16606145 FC204 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16686135 Supplier Gilson™ Supplier No. 170730
  • ONLINE EXCLUSIVE PRICE
    8895.00 DKK
    7484.58 DKK / Pakke med 1
    Spare 1410.42 DKK
    15% Slukket
Forventet forsendelse: 05-01-2027
These assemblies and packages add fraction collection capabilities to various Gilson instruments.
  • Purification Technique HPLC
  • Used With Fraction Collector
Specifications
Description FC203
Diameter (Metric) Inner 1.5 mm