missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
3-Tropanyl-3,5-dichlorobenzoate, 99+%, Thermo Scientific™
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is most often used to study the role of SR-3. Studies suggest the blockade of SR-3 with may ameliorate beta-amyloid protein-induced neurotoxicity by interfering with the increase of Ca2+, and then by inhibiting glutamate release,
It is most often used to study the role of SR-3. Studies suggest the blockade of SR-3 with may ameliorate beta-amyloid protein-induced neurotoxicity by interfering with the increase of Ca2+, and then by inhibiting glutamate release,
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Tropanyl-3,5-dichlorobenzoate |
| CAS | 40796-97-2 |
| Farve | White |
| Molekylær formel | C15H17Cl2NO2 |
| MDL nummer | RONG> |
| Mængde | 50 mg |
| Synonym | bemesetron, bemesetronum, unii-o98t3677pa, tropyl 3,5-dichlorobenzoate, bemesetron usan:inn, bemesetronum inn-latin, dsstox_cid_22632, dsstox_rid_80063, dsstox_gsid_42632, tropanyl 3,5-dichlorobenzoate |
| Opløselighedsinformation | Soluble to 100 mM in DMSO. Insoluble in water. Sparingly soluble in chloroform,and ethanol. |
| InChI nøgle | MNJNPLVXBISNSX-PBWFPOADSA-N |
| SMIL | CN1C2CCC1CC(C2)OC(=O)C3=CC(=CC(=C3)Cl)Cl |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?