Få mere at vide
3-Trimethylstannyl Benzoic Acid, TRC
Artikelnummer. 30454159
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454159 10 mg
30454157 5 mg
30456757 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30454159 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79719010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 161767-56-2
Molekylær formel C10 H14 O2 Sn
Molekylvægt (g/mol) 284.93
IUPAC navn 3-trimethylstannylbenzoic acid
SMIL C[Sn](C)(C)c1cccc(c1)C(=O)O
Tekniske data
Kemisk navn eller materiale 3-Trimethylstannyl Benzoic Acid
Mængde 10 mg
InChI formel InChI=1S/C7H5O2.3CH3.Sn/c8-7(9)6-4-2-1-3-5-6;;;;/h1-2,4-5H,(H,8,9);3*1H3;
Formel vægt 286.0016