missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilylpropynal, 97%, Thermo Scientific Chemicals

Catalog Number p-7059518
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
2975-46-4
C6H10OSi
126.23
MFCD01103709
LJRWLSKYGWLYIM-UHFFFAOYSA-N
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
2760865
3-trimethylsilylprop-2-ynal
C[Si](C)(C)C#CC=O
This item is not returnable. View return policy
2975-46-4
C6H10OSi
126.23
MFCD01103709
LJRWLSKYGWLYIM-UHFFFAOYSA-N
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
2760865
3-trimethylsilylprop-2-ynal
C[Si](C)(C)C#CC=O

3-Trimethylsilylpropynal acts as an important organic intermediate. It is used in agrochemicals, pharmaceuticals and dyestuff field. Further, it is used in the preparation of [3-(trimethylsilyl)-2-propinyliden]-malononitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.86g/mL
Boiling Point 58°C to 60°C (37.5 mmHg)
Flash Point 29°C (84°F)
Linear Formula (CH3)3SiC≡CCHO
Refractive Index 1.4435
Quantity 0.25 g
UN Number UN1993
Sensitivity Air & Moisture Sensitive
Formula Weight 126.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilylpropynal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'