missing translation for 'onlineSavingsMsg'
Learn More

3-[(Trimethylsilyl)ethynyl]pyridine, 97%, Alfa Aesar™

Brand:  Alfa Aesar H55101.06

Additional Details : CAS Number : 80673-00-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15471248

  • 565.31 DKK / 5g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

80673-00-3
175.306
MTMBLPOPETZZRX-UHFFFAOYSA-N
4100857
C[Si](C)(C)C#CC1=CN=CC=C1
C10H13NSi
MFCD01863744
3-trimethylsilyl ethynyl pyridine, 3-trimethylsilylethynyl pyridine, 3-pyridylethynyl trimethylsilane, 3-2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-3-ylethynyl silane, acmc-209pjw, 3-trimethylsilylethynylpyridine, trimethyl-2-3-pyridyl ethynyl silane, 1-3-pyridyl-2-trimethylsilylacetylene
trimethyl(2-pyridin-3-ylethynyl)silane

Specifications

3-[(Trimethylsilyl)ethynyl]pyridine
94°C (201°F)
229°C
5g
C10H13NSi
3-trimethylsilyl ethynyl pyridine, 3-trimethylsilylethynyl pyridine, 3-pyridylethynyl trimethylsilane, 3-2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-3-ylethynyl silane, acmc-209pjw, 3-trimethylsilylethynylpyridine, trimethyl-2-3-pyridyl ethynyl silane, 1-3-pyridyl-2-trimethylsilylacetylene
C[Si](C)(C)C#CC1=CN=CC=C1
175.306
175.31
97%
0.922 g/mL
1.529
Colorless to Yellow
80673-00-3
MFCD01863744
MTMBLPOPETZZRX-UHFFFAOYSA-N
trimethyl(2-pyridin-3-ylethynyl)silane
4100857
Liquid
Videos
SDS Documents