Learn More
3-(Trifluoromethyl)phenylacetic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 351-35-9 |
| Molecular Formula | C9H7F3O2 |
| MDL Number | MFCD00004339 |
| Quantity | 100 g |
| SMILES | O=C(O)CC1=CC=CC(C(F)(F)F)=C1 |
| Molecular Weight (g/mol) | 204.15 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-(Trifluoromethyl)phenylacetic acid |