missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenol, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020727
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
98-17-9
C7H5F3O
162.111
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F
This item is not returnable. View return policy
98-17-9
C7H5F3O
162.111
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2°C
Density 1.336
Boiling Point 177°C to 178°C
Flash Point 74°C (165°F)
Odor Phenol-like
Refractive Index 1.458
Quantity 5 g
UN Number UN3265
Beilstein 2045663
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'