Få mere at vide
3-Trifluoromethyl Hydroxychloroquine, TRC
Artikelnummer. 30451736
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30451736 10 mg -
30449957 25 mg -
30449256 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30451736 Leverandør Toronto Research Chemicals Leverandørnr. TRCT81584010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2054858-88-5
Molekylær formel C19H25ClF3N3O
Molekylvægt (g/mol) 403.87
Synonym 2-((4-((7-Chloro-3-(trifluoromethyl)quinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol
SMIL CCN(CCO)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1C(F)(F)F
Tekniske data
Kemisk navn eller materiale 3-Trifluoromethyl Hydroxychloroquine
Mængde 10 mg
InChI formel InChI=1S/C19H25ClF3N3O/c1-3-26(9-10-27)8-4-5-13(2)25-18-15-7-6-14(20)11-17(15)24-12-16(18)19(21,22)23/h6-7,11-13,27H,3-5,8-10H2,1-2H3,(H,24,25)
Formel vægt 403.1638