Få mere at vide
[3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol, TRC
Artikelnummer. 30456005
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456005 250 mg
30453030 25 mg
30452258 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456005 Leverandør Toronto Research Chemicals Leverandørnr. TRCT898930250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 169213-73-4
Molekylær formel C5H5N2OF3
Molekylvægt (g/mol) 166.1
Synonym 5-(Trifluoromethyl)-1H-pyrazole-3-methanol, (5-Trifluoromethyl-2H-pyrazol-3-yl)-methanol, [5-(Trifluoromethyl)-1H-pyrazol-3-yl]methanol
IUPAC navn [3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol
SMIL OCc1cc(n[nH]1)C(F)(F)F
Tekniske data
Kemisk navn eller materiale [3-(trifluoromethyl)-1H-pyrazol-5-yl]methanol
Mængde 250 mg
InChI formel InChI=1S/C5H5F3N2O/c6-5(7,8)4-1-3(2-11)9-10-4/h1,11H,2H2,(H,9,10)
Formel vægt 166.0354