missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Catalog Number p-205114
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1535-73-5
MFCD00041511
SADHVOSOZBAAGL-UHFFFAOYSA-N
3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
73753
3-(trifluoromethoxy)aniline
C1=CC(=CC(=C1)OC(F)(F)F)N
This item is not returnable. View return policy
1535-73-5
MFCD00041511
SADHVOSOZBAAGL-UHFFFAOYSA-N
3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
73753
3-(trifluoromethoxy)aniline
C1=CC(=CC(=C1)OC(F)(F)F)N
CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.3200g/mL
Boiling Point 72.0°C to 75.0°C (8.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H6F3NO
Linear Formula CF3OC6H4NH2
Refractive Index 1.4650 to 1.4670
Quantity 500 mg
Specific Gravity 1.32
Molecular Weight (g/mol) 177.12
Formula Weight 177.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)aniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)aniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'