Få mere at vide
3-Toluoyl Decitabine, TRC
Artikelnummer. 30456108
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456108 100 mg
30449060 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456108 Leverandør Toronto Research Chemicals Leverandørnr. TRCT536260100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 183016-21-9
Molekylær formel C16H18N4O5
Molekylvægt (g/mol) 346.34
Synonym 4-Amino-1-[2-deoxy-3-O-(4-methylbenzoyl)-β-D-erythro-pentofuranosyl]-1,3,5-triazin-2(1H)-one
IUPAC navn [(2R,3S,5R)-5-(4-amino-2-oxo-1,3,5-triazin-1-yl)-2-(hydroxymethyl)oxolan-3-yl] 4-methylbenzoate
SMIL Cc1ccc(cc1)C(=O)O[C@H]2C[C@@H](O[C@@H]2CO)N3C=NC(=NC3=O)N
Tekniske data
Kemisk navn eller materiale 3-Toluoyl Decitabine
Mængde 100 mg
InChI formel InChI=1S/C16H18N4O5/c1-9-2-4-10(5-3-9)14(22)25-11-6-13(24-12(11)7-21)20-8-18-15(17)19-16(20)23/h2-5,8,11-13,21H,6-7H2,1H3,(H2,17,19,23)/t11-,12+,13+/m0/s1
Formel vægt 346.1277