Få mere at vide
3-Sulfoglycolithocholic Acid Disodium Salt, TRC
Artikelnummer. 30137205
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137205 50 mg
30005369 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137205 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68911550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64936-82-9
Molekylær formel C26 H41 N O7 S . 2 Na
Molekylvægt (g/mol) 557.65
Synonym N-[(3α,5β)-24-Oxo-3-(sulfooxy)cholan-24-yl]glycine Disodium Salt
IUPAC navn disodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-sulfonatooxy-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
SMIL [Na+].[Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale 3-Sulfoglycolithocholic Acid Disodium Salt
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C26H43NO7S.2Na/c1-16(4-9-23(28)27-15-24(29)30)20-7-8-21-19-6-5-17-14-18(34-35(31,32)33)10-12-25(17,2)22(19)11-13-26(20,21)3;;/h16-22H,4-15H2,1-3H3,(H,27,28)(H,29,30)(H,31,32,33);;/q;2*+1/p-2/t16-,17-,18-,19+,20-,21+,22+,25+,26-;;/m1../s1
Formel vægt 557.2399
Procent renhed >95%