Få mere at vide
(3’R)-Ezetimibe, TRC
Artikelnummer. 17974285
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974285 25 mg
17941285 10 mg
17931285 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17974285 Leverandør Toronto Research Chemicals Leverandørnr. TRCE97501025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163380-16-3
Molekylær formel C24 H21 F2 N O3
Molekylvægt (g/mol) 409.43
Synonym (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-, [3R-[3α(R*),4β]]-, 3'R,3R,4S-Isomer of Ezetimibe, R,R,S-Ezetimibe, (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone, R,R,S-Ezetimibe, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-, [3R-[3alpha(R*),4beta]]-, 3’R,3R,4S-Isomer of Ezetimibe
IUPAC navn (3R,4S)-1-(4-fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one
SMIL O[C@H](CC[C@@H]1[C@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccc(F)cc4
Tekniske data
Kemisk navn eller materiale (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C24H21F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21-23,28-29H,13-14H2/t21-,22-,23-/m1/s1
Formel vægt 409.1489
Procent renhed >95%