Få mere at vide
3-Quinuclidinol-d7, TRC
Artikelnummer. 30422376
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30422376 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422376 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ79578225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Quinuclidinol-d7
CAS 2708283-05-8
Molekylær formel C7 D7 H6 N O
Mængde 25 mg
Synonym 2,2,3,6,6,7,7-Heptadeuterioquinuclidin-3-ol, 3-Quinuclidinol-d7, 3-Quinuclidinol-2,2,3,6,6,7,7-D7, 2,2,3,6,6,7,7-Heptadeuterio-1-azabicyclo[2.2.2]octan-3-ol, 1-Azabicyclo[2.2.2]octan-3-ol-2,2,3,6,6,7,7-d7, (±)-3-Quinuclidinol-2,2,3,6,6,7,7-d7
InChI formel InChI=1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2/i3D2,4D2,5D2,7D
SMIL [2H]C1([2H])CC2CC([2H])([2H])N1C([2H])([2H])C2([2H])O
IUPAC navn 2,2,3,6,6,7,7-heptadeuterio-1-azabicyclo[2.2.2]octan-3-ol
Molekylvægt (g/mol) 134.2274
Formel vægt 134.1437