Få mere at vide
(±)-3-Quinuclidinol-2,2,3,6,6,7,7-d7, CDN
Artikelnummer. 30321942
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321942 0.05 g -
30311561 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321942 Leverandør CDN Leverandørnr. CDND75600.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Quinuclidinol-d7
CAS 2708283-05-8
Molekylær formel C7 D7 H6 N O
Mængde 0.05 g
Synonym 2,2,3,6,6,7,7-Heptadeuterioquinuclidin-3-ol, 3-Quinuclidinol-d7, 3-Quinuclidinol-2,2,3,6,6,7,7-D7, 2,2,3,6,6,7,7-Heptadeuterio-1-azabicyclo[2.2.2 ]octan-3-ol, 1-Azabicyclo[2.2.2 ]octan-3-ol-2,2,3,6,6,7,7-d7, (±)-3-Quinuclidinol-2,2,3,6,6,7,7-d7
InChI formel InChI=1 S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9 H,1-5H2/i3D2,4D2,5D2,7 D
SMIL [2 H]C1([2 H])CC2CC([2 H])([2 H])N1C([2 H])([2 H])C2([2 H])O
IUPAC navn 2,2,3,6,6,7,7-heptadeuterio-1-azabicyclo[2.2.2 ]octan-3-ol
Molekylvægt (g/mol) 134.2274
Formel vægt 134.1437 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity