missing translation for 'onlineSavingsMsg'
Learn More

3-(quinoxalin-2-yl)prop-2-enoic Acid, TRC

Product Code. 30385562
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30385562 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385562 Supplier Toronto Research Chemicals Supplier No. TRCE59244025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-(quinoxalin-2-yl)prop-2-enoic Acid
CAS 1593-24-4
Molekylær formel C11 H8 N2 O2
Mængde 25 mg
Synonym 3-(2-Quinoxalinyl)-2-propenoic acid, NSC 158311
InChI formel InChI=1S/C11H8N2O2/c14-11(15)6-5-8-7-12-9-3-1-2-4-10(9)13-8/h1-7H,(H,14,15)/b6-5+
SMIL OC(=O)\C=C\c1cnc2ccccc2n1
IUPAC navn (E)-3-quinoxalin-2-ylprop-2-enoic acid
Molekylvægt (g/mol) 200.193
Formel vægt 200.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.