Få mere at vide
3-Pyridylcarbinol-d6, TRC
Artikelnummer. 30414171
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30414171 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414171 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99280225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Pyridylcarbinol-d6
CAS 1189493-62-6
Molekylær formel C6HD6NO
Mængde 25 mg
Synonym Dideuterio-(2,4,5,6-tetradeuteriopyridin-3-yl)methanol, 3-Pyridinemethanol-d6, 3-Pyridinemethanol-α,α,2,4,5,6-d6, 3-(Hydroxymethyl)pyridine-D6, 3-(Hydroxymethyl-d2)pyridine-2,4,5,6-d4, Nicotinic alcohol-d6, Nicotinyl alcohol-d6, β-Picolyl alcohol-d6
InChI formel InChI=1S/C6H7NO/c8-5-6-2-1-3-7-4-6/h1-4,8H,5H2/i1D,2D,3D,4D,5D2
SMIL [2H]c1nc([2H])c(c([2H])c1[2H])C([2H])([2H])O
IUPAC navn dideuterio-(2,4,5,6-tetradeuteriopyridin-3-yl)methanol
Molekylvægt (g/mol) 115.16
Formel vægt 115.0904